Bromoacetaldehyde diethyl acetal ≥95.0%

Supplier: TCI America

Synonyms: 2-Bromo-1,1-diethoxyethane

B0957-500ML B0957-25ML B0957-100ML
TCB0957-500MLEA 310 USD
TCB0957-500ML TCB0957-25ML TCB0957-100ML
Bromoacetaldehyde diethyl acetal ≥95.0%
Bromoacetaldehyde diethyl acetal

CAS Number: 2032-35-1
MDL Number: MFCD00000214
Molecular Formula: C6H13BrO2
Molecular Weight: 197.07
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 67
Flash Point (°C): 51
Specific Gravity (20/20): 1.28


Other Product Names / Synonyms: 2-Bromo-1,1-diethoxyethane, 2,2-Diethoxyethyl Bromide

Formula: BrH₂CCH(OCH₂CH₃)₂
Boiling Pt: 67 °C
Density: 1.28 (20 °C)
Flash Pt: 51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000214
CAS Number: 2032-35-1
EINECS: 217-989-1
UN: 2810
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=082a4a331b1e17b9849f92ed506aff2762ca5fd0&vid=b1974fde73c4f6f71fa7926d886148863bc44c23&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR