2-Bromothiophene ≥98.0%
Supplier: TCI America
Synonyms:
2-Thienyl bromide
B0941-250G
B0941-25G
TCB0941-250GEA
132
USD
TCB0941-250G
TCB0941-025G
2-Bromothiophene ≥98.0%
2-Bromothiophene
CAS Number: 1003-09-4
MDL Number: MFCD00005417
Molecular Formula: C4H3BrS
Molecular Weight: 163.03
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 154
Flash Point (°C): 60
Specific Gravity (20/20): 1.70
Other Product Names / Synonyms: 2-Thienyl Bromide
Formula:
C₄H₃BrS MW: 163.04 g/mol Boiling Pt: 154 °C Melting Pt: –10 °C Density: 1.70 (20 °C) Flash Pt: 60 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005417 CAS Number: 1003-09-4 EINECS: 213-699-4 UN: 2929 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...