4-Bromoisoquinoline ≥98.0%
Supplier: TCI America
|
Warning
|
B0930-10G
B0930-25G
TCB0930-010GEA
70
USD
TCB0930-010G
TCB0930-025G
4-Bromoisoquinoline ≥98.0%
4-Bromoisoquinoline
CAS Number: 1532-97-4
MDL Number: MFCD00006904
Molecular Formula: C9H6BrN
Molecular Weight: 208.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 285
Melting point (°C): 42
Flash Point (°C): 110
Formula:
C₉H₆BrN MW: 208.06 g/mol Boiling Pt: 285 °C Melting Pt: 42 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006904 CAS Number: 1532-97-4 EINECS: 216-244-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...