2-tert-Butyl-p-cresol ≥98.0%

Supplier: TCI America
Warning

Synonyms: 1-tert-Butyl-2-hydroxy-5-methylbenzene, 2-tert-Butyl-4-methylphenol, 3-tert-Butyl-4-hydroxytoluene

B0910-500G B0910-25G
TCB0910-500GEA 74 USD
TCB0910-500G TCB0910-25G
2-tert-Butyl-p-cresol ≥98.0%
2-tert-Butyl-p-cresol

CAS Number: 2409-55-4
MDL Number: MFCD00002381
Molecular Formula: C11H16O
Molecular Weight: 164.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 244
Melting point (°C): 52
Flash Point (°C): 47


Other Product Names / Synonyms: 2-tert-Butyl-4-methylphenol

Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Boiling Pt: 244 °C
Melting Pt: 52 °C
Density: 0.961 g/cm³ (20 °C)
Flash Pt: 47 °C
Storage Temperature: Ambient
MDL Number: MFCD00002381
CAS Number: 2409-55-4
EINECS: 219-314-6
UN: 2430
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=053c85f9d90e4199125c2369a142adea3a0f53b5&vid=053c85f9d90e4199125c2369a142adea3a0f53b5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR