1-Bromo-2-fluorobenzene ≥99.0%
Supplier: TCI America
CAS Number: 1072-85-1
MDL Number: MFCD00000282
Molecular Formula: C6H4BrF
Molecular Weight: 175.00
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Melting point (°C): -8
Flash Point (°C): 48
Specific Gravity (20/20): 1.61
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 1-Bromo-2-fluorobenzene
Formula:
C₆H₄BrF Boiling Pt: 158 °C Melting Pt: –8 °C Density: 1.61 (20 °C) Flash Pt: 48 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000282 CAS Number: 1072-85-1 EINECS: 214-018-3 UN: 1993 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...