3-Benzoylpropionic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Oxo-4-phenylbutanoic acid, 3-Benzoylpropionicacid

B0868-25G B0868-500G
TCB0868-025GEA 79 USD
TCB0868-025G TCB0868-500G
3-Benzoylpropionic acid ≥98.0% (by GC, titration analysis)
3-Benzoylpropionic acid

CAS Number: 2051-95-8
MDL Number: MFCD00002792
Molecular Formula: C10H10O3
Molecular Weight: 178.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 118


Other Product Names / Synonyms: 4-Oxo-4-phenylbutyric Acid

Formula: C₁₀H₁₀O₃
MW: 178.19 g/mol
Boiling Pt: 325 °C (1013 hPa)
Melting Pt: 118 °C
MDL Number: MFCD00002792
CAS Number: 2051-95-8
EINECS: 218-135-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=88c5c267568982dda021d0434ca70454345adfdd&vid=88c5c267568982dda021d0434ca70454345adfdd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR