2-Bromo-4,6-dinitroaniline ≥98.0%

Supplier: TCI America

B0814-500G B0814-25G
TCB0814-500GEA 131 USD
TCB0814-500G TCB0814-25G
2-Bromo-4,6-dinitroaniline ≥98.0%
2-Bromo-4,6-dinitroaniline

CAS Number: 1817-73-8
MDL Number: MFCD00007146
Molecular Formula: C6H4BrN3O4
Molecular Weight: 262.02
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 152

Formula: C₆H₄BrN₃O₄
MW: 262.02 g/mol
Boiling Pt: 401 °C (1013 hPa)
Melting Pt: 152 °C
MDL Number: MFCD00007146
CAS Number: 1817-73-8
EINECS: 217-329-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db4496bf2fbf1234d0d2278b1ca261fabd62d2c3&vid=0eaf60621148852c8c5becc9ce2cdecb68a3557e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR