4-Bromophenol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

B0787-500G B0787-25G B0787-100G
TCB0787-500GEA 115.18 USD
TCB0787-500G TCB0787-25G TCB0787-100G
4-Bromophenol ≥98.0% (by GC, titration analysis)
4-Bromophenol

CAS Number: 106-41-2
MDL Number: MFCD00002313
Molecular Formula: C6H5BrO
Molecular Weight: 173.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 238
Melting point (°C): 65
Flash Point (°C): 241
Lambda max.: 282 nm (EtOH)

Formula: C₆H₅BrO
MW: 173.01 g/mol
Boiling Pt: 238 °C
Melting Pt: 65 °C
Density: ~ 1.84 g/cm³ (20 °C)
Flash Pt: 241 °C
Storage Temperature: Ambient
MDL Number: MFCD00002313
CAS Number: 106-41-2
EINECS: 203-394-4
UN: 2811
ADR: 6.1,III
Merck Index: 13,01413
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c07e7e461be5aabf9bb645a748408a13a66d81ec&vid=53dd3c9344ec7e8d27e545283fa3c4178c325b01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR