Isoamyl butyrate ≥98.0%
Supplier: TCI America
Synonyms:
Isopentyl butyrate, Isoamyl Butyrate
B0760-500ML
B0760-25ML
TCB0760-500MLEA
54
USD
TCB0760-500ML
TCB0760-25ML
Isoamyl butyrate ≥98.0%
Isoamyl butyrate
CAS Number: 106-27-4
MDL Number: MFCD00044888
Molecular Formula: C9H18O2
Molecular Weight: 158.24
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 179
Flash Point (°C): 65
Specific Gravity (20/20): 0.87
Other Product Names / Synonyms: Butyric Acid Isoamyl Ester
Formula:
C₉H₁₈O₂ MW: 158.24 g/mol Boiling Pt: 179 °C Density: 0.87 (20 °C) Flash Pt: 65 °C |
MDL Number:
MFCD00044888 CAS Number: 106-27-4 EINECS: 203-380-8 UN: 2620 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...