Ethyl butyrate ≥98.0%

Supplier: TCI America

Synonyms: Butanoic acid ethyl ester, Butyric acid ethyl ester

B0759-500ML B0759-25ML
TCB0759-500MLEA 29.3 USD
TCB0759-500ML TCB0759-25ML
Ethyl butyrate ≥98.0%
Ethyl butyrate

CAS Number: 105-54-4
MDL Number: MFCD00009394
Molecular Formula: C6H12O2
Molecular Weight: 116.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 23
Specific Gravity (20/20): 0.88


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Butyric Acid Ethyl Ester

Formula: C₆H₁₂O₂
Boiling Pt: 120 °C
Melting Pt: –92 °C
Density: 0.88 (20 °C)
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00009394
CAS Number: 105-54-4
EINECS: 203-306-4
UN: 1180
ADR: 3,III
Merck Index: 13,03810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c07d6f7c3c418dbf15513ac4993e399e5ef52d04&vid=c07d6f7c3c418dbf15513ac4993e399e5ef52d04&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR