Butyl butyrate ≥99.0%

Supplier: TCI America
Danger

Synonyms: Butyric acid butyl ester

B0757-500ML B0757-25ML
TCB0757-500MLEA 57.14 USD
TCB0757-500ML TCB0757-025ML
Butyl butyrate ≥99.0%
Butyl butyrate

CAS Number: 109-21-7
MDL Number: MFCD00009450
Molecular Formula: C8H16O2
Molecular Weight: 144.21
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 164
Melting point (°C): -92
Flash Point (°C): 56
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Butyric Acid Butyl Ester

Formula: C₈H₁₆O₂
MW: 144.21 g/mol
Boiling Pt: 164 °C
Melting Pt: -92 °C
Density: 0.87 (20 °C)
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD00009450
CAS Number: 109-21-7
EINECS: 203-656-8
UN: 3272
ADR: 3,III
Merck Index: 13,01555
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e8f4f01187c3b7f299a5fa762c6310e9047d316c&vid=e8f4f01187c3b7f299a5fa762c6310e9047d316c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR