Butyraldehyde oxime ≥95.0%
Supplier: TCI America
Synonyms:
Butyraldoxime
B0753-500ML
B0753-25ML
TCB0753-500MLEA
846
USD
TCB0753-500ML
TCB0753-25ML
Butyraldehyde oxime ≥95.0%
Butyraldehyde oxime
CAS Number: 110-69-0
MDL Number: MFCD00013943
Molecular Formula: C4H9NO
Molecular Weight: 87.12
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 152
Flash Point (°C): 58
Specific Gravity (20/20): 0.91
Other Product Names / Synonyms: Butyraldehyde Oxime
Formula:
C₄H₉NO Boiling Pt: 152 °C Density: 0.91 (20 °C) Flash Pt: 58 °C |
MDL Number:
MFCD00013943 CAS Number: 110-69-0 EINECS: 203-792-8 UN: 2840 ADR: 3,III |
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...