2-Butyne-1,4-diol ≥99.0%
Supplier: TCI America
|
Danger
|
Synonyms:
1,4-Dihydroxy-2-butyne, But-2-yne-1,4-diol
B0749-500G
B0749-25G
TCB0749-500GEA
77.56
USD
TCB0749-500G
TCB0749-25G
2-Butyne-1,4-diol ≥99.0%
2-Butyne-1,4-diol
CAS Number: 110-65-6
MDL Number: MFCD00002915
Molecular Formula: C4H6O2
Molecular Weight: 86.09
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 238
Melting point (°C): 58
Flash Point (°C): 128
Other Product Names / Synonyms: 1,4-Dihydroxy-2-butyne, 1,2-Dihydroxydimethylacetylene
Formula:
C₄H₆O₂ MW: 86.09 g/mol Boiling Pt: 238 Melting Pt: 58 °C Density: 1.1001 g/cm³ (20 °C) Flash Pt: 128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002915 CAS Number: 110-65-6 EINECS: 203-788-6 UN: 2716 ADR: 6.1 III |
|
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...