Butyl vinyl ether ≥98.0% stabilized
Supplier: TCI America
Synonyms:
n-Butyl vinyl ether
Stabilization:
Stabilised with Stabilised
B0746-500ML
B0746-25ML
TCB0746-500MLEA
58.19
USD
TCB0746-500ML
TCB0746-25ML
Butyl vinyl ether ≥98.0% stabilized
Butyl vinyl ether
(stabilized with KOH)
CAS Number: 111-34-2
MDL Number: MFCD00009454
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Melting point (°C): -92
Flash Point (°C): -9
Specific Gravity (20/20): 0.78
Storage Temperature: 0-10°C
Formula:
C₆H₁₂O MW: 100.16 g/mol Boiling Pt: 93 °C Melting Pt: -92 °C Density: 0.78 (20 °C) Flash Pt: -9 °C |
MDL Number:
MFCD00009454 CAS Number: 111-34-2 EINECS: 203-860-7 UN: 3271 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...