2,2'-Butyliminodiethanol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
CAS Number: 102-79-4
MDL Number: MFCD00002856
Molecular Formula: C8H19NO2
Molecular Weight: 161.25
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 275
Melting point (°C): -70
Flash Point (°C): 143
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: 2,2'-(Butylimino)diethanol, N,N-Bis(2-hydroxyethyl)butylamine
Formula:
H₃CCH₂CH₂CH₂N(CH₂CH₂OH)₂ Boiling Pt: 275 °C Melting Pt: –70 °C Density: 0.97 (20 °C) Flash Pt: 143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002856 CAS Number: 102-79-4 EINECS: 203-055-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...