Dibutyl ether ≥99.0% stabilized
Supplier: TCI America
|
Danger
|
Synonyms:
Di-n-butyl ether
Stabilization:
Stabilised with Stabilised
B0721-500ML
B0721-25ML
TCB0721-500MLEA
40.66
USD
TCB0721-500ML
TCB0721-25ML
Dibutyl ether ≥99.0% stabilized
Dibutyl ether
(stabilized with BHT)
CAS Number: 142-96-1
MDL Number: MFCD00009461
Molecular Formula: C8H18O
Molecular Weight: 130.23
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 141
Melting point (°C): -98
Flash Point (°C): 37
Specific Gravity (20/20): 0.77
Other Product Names / Synonyms: Butyl Ether
Formula:
C₈H₁₈O MW: 130.23 g/mol Boiling Pt: 141 °C Melting Pt: –98 °C Density: 0.77 (20 °C) Flash Pt: 37 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00009461 CAS Number: 142-96-1 EINECS: 205-575-3 UN: 1149 ADR: 3,III Merck Index: 13,01568 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...