(±)-sec-Butylamine ≥99.0% (by GC, titration analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
(±)-2-Butylamine, (±)-2-Aminobutane
B0708-500ML
B0708-25ML
TCB0708-500MLEA
53
USD
TCB0708-500ML
TCB0708-25ML
(±)-sec-Butylamine ≥99.0% (by GC, titration analysis)
(±)-sec-Butylamine
CAS Number: 13952-84-6
MDL Number: MFCD00008094
Molecular Formula: C4H11N
Molecular Weight: 73.14
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 63
Flash Point (°C): -9
Specific Gravity (20/20): 0.73
Other Product Names / Synonyms: 2-Aminobutane
Formula:
C₄H₁₁N MW: 73.14 g/mol Boiling Pt: 63 °C Melting Pt: –104 °C Density: 0.73 (20 °C) Flash Pt: –9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008094 CAS Number: 13952-84-6 EINECS: 237-732-7 UN: 2733 ADR: 3,II Merck Index: 12,01579 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...