Butylamine ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 1-Butylamine, 1-Aminobutane, n-Butylamine

B0707-500ML B0707-25ML
TCB0707-500MLEA 45.8 USD
TCB0707-500ML TCB0707-25ML
Butylamine ≥99.0% (by GC, titration analysis)
Butylamine

CAS Number: 109-73-9
MDL Number: MFCD00011690
Molecular Formula: C4H11N
Molecular Weight: 73.14
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 77
Melting point (°C): -49
Flash Point (°C): -4
Specific Gravity (20/20): 0.74


Product Categories: Synthetic Organic Chemistry, Amines (Multi-component Reaction), Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1-Aminobutane

Formula: H₃C(CH₂)₂CH₂NH₂
MW: 73.14 g/mol
Boiling Pt: 77 °C
Melting Pt: -49 °C
Density: 0.74 (20 °C)
Flash Pt: -4 °C
Storage Temperature: Ambient
MDL Number: MFCD00011690
CAS Number: 109-73-9
EINECS: 203-699-2
UN: 1125
ADR: 3,II
Merck Index: 12,01578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9172c8538459af4a13da3451a3a89dd14ef33ee&vid=223e471a5e49594cf78481996f2204ac717c014d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR