2-Butoxyethanol ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
Ethylene glycol butyl ether, Butyl cellosolve, Ethylene glycol monobutyl ether
B0698-500ML
B0698-25ML
TCB0698-500MLEA
36.74
USD
TCB0698-500ML
TCB0698-25ML
2-Butoxyethanol ≥99.0%
2-Butoxyethanol
CAS Number: 111-76-2
MDL Number: MFCD00002884
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 171
Melting point (°C): -75
Flash Point (°C): 68
Specific Gravity (20/20): 0.90
Product Categories: Materials Chemistry, Polyimide Raw Materials
Other Product Names / Synonyms: 2-Butoxyethanol, Butyl Cellosolve, Butyl Glycol
Formula:
CH₃(CH₂)₃OCH₂CH₂OH MW: 118.18 g/mol Boiling Pt: 171 °C Melting Pt: –75 °C Density: 0.90 (20 °C) Flash Pt: 68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002884 CAS Number: 111-76-2 EINECS: 203-905-0 UN: 2810 ADR: 6.1,III Merck Index: 13,01558 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...