1-Butanethiol ≥97.0%
Supplier: TCI America
Synonyms:
1-Mercaptobutane, n-Butanethiol, NBM, n-Butyl mercaptan
B0685-500ML
B0685-25ML
TCB0685-500MLEA
53
USD
TCB0685-500ML
TCB0685-25ML
1-Butanethiol ≥97.0%
1-Butanethiol
CAS Number: 109-79-5
MDL Number: MFCD00004905
Molecular Formula: C4H10S
Molecular Weight: 90.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 2
Specific Gravity (20/20): 0.84
Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Butyl Mercaptan
Formula:
C₄H₁₀S MW: 90.19 g/mol Boiling Pt: 98 °C Melting Pt: –116 °C Density: 0.84 (20 °C) Flash Pt: 2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004905 CAS Number: 109-79-5 EINECS: 203-705-3 UN: 2347 ADR: 3,II Merck Index: 12,01611 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...