Diacetyl ≥98.0%

Supplier: TCI America
Danger

Synonyms: 2,3-Butanedione

B0682-500ML B0682-25ML B0682-100ML
TCB0682-500MLEA 103.53 USD
TCB0682-500ML TCB0682-25ML TCB0682-100ML
Diacetyl ≥98.0%
Diacetyl

CAS Number: 431-03-8
MDL Number: MFCD00008756
Molecular Formula: C4H6O2
Molecular Weight: 86.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Melting point (°C): -4
Flash Point (°C): 11
Specific Gravity (20/20): 0.98
Storage Temperature: 0-10°C


Product Categories: Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: 2,3-Butanedione, 2,3-Diketobutane, Dimethylglyoxal

Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 88 °C
Melting Pt: –4 °C
Density: 0.98 (20 °C)
Flash Pt: 11 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008756
CAS Number: 431-03-8
EINECS: 207-069-8
UN: 2346
ADR: 3,II
Merck Index: 13,02985
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=400243065b1b026f80284d72df81a61550d4fda2&vid=efa2d3b18bb5cdcefd3b4d9ff5a4d2aba7467728&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR