4-Bromotoluene ≥99.0%

Supplier: TCI America

B0661-25G B0661-500G B0661-100G
TCB0661-025GEA 52.72 USD
TCB0661-025G TCB0661-500G TCB0661-100G
4-Bromotoluene ≥99.0%
4-Bromotoluene

CAS Number: 106-38-7
MDL Number: MFCD00000109
Molecular Formula: C7H7Br
Molecular Weight: 171.04
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 184
Flash Point (°C): 85
Freezing point (°C): 26

Formula: C₇H₇Br
Boiling Pt: 184 °C
Melting Pt: 24…27 °C
Density: 1.387 g/cm³ (20 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00000109
CAS Number: 106-38-7
EINECS: 203-391-8
UN: 3077
ADR: 9,III
Merck Index: 13,01424
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bfff899c2ae74501b4ba8a41c556ba48f882259&vid=0382cf87239f26e08163d63226685b14caceb70f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR