(±)-2-Bromopropionyl bromide ≥98.0%

Supplier: TCI America

B0649-500G B0649-25G B0649-100G
TCB0649-500GEA 154 USD
TCB0649-500G TCB0649-25G TCB0649-100G
(±)-2-Bromopropionyl bromide ≥98.0%
(±)-2-Bromopropionyl bromide

CAS Number: 563-76-8
MDL Number: MFCD00000142
Molecular Formula: C3H4Br2O
Molecular Weight: 215.87
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 155
Flash Point (°C): 109
Specific Gravity (20/20): 2.05

Formula: C₃H₄Br₂O
MW: 215.87 g/mol
Boiling Pt: 155 °C
Density: 2.05 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00000142
CAS Number: 563-76-8
EINECS: 209-261-7
UN: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66d8aa5fc8efa31db63b13ca561bb0a018764619&vid=66d8aa5fc8efa31db63b13ca561bb0a018764619&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR