Allyl bromide ≥98.0%
Supplier: TCI America
Synonyms:
3-Bromopropene, 3-Propenyl bromide
CAS Number: 106-95-6
MDL Number: MFCD00000244
Molecular Formula: C3H5Br
Molecular Weight: 120.98
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 71
Flash Point (°C): 12
Specific Gravity (20/20): 1.43
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Materials Chemistry, Precise Polymerization Reagents, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: 3-Bromo-1-propene, 3-Bromopropylene
Formula:
C₃H₅Br MW: 120.98 g/mol Boiling Pt: 70 Melting Pt: -119 Density: 1.43 (20 °C) Flash Pt: 12 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000244 CAS Number: 106-95-6 EINECS: 203-446-6 UN: 1099 ADR: 3 I Merck Index: 13,00285 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...