1-Bromopropane ≥98.0%
Supplier: TCI America
|
Danger
|
CAS Number: 106-94-5
MDL Number: MFCD00000254
Molecular Formula: C3H7Br
Molecular Weight: 122.99
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 71
Melting point (°C): -110
Flash Point (°C): 25
Specific Gravity (20/20): 1.36
Other Product Names / Synonyms: Propyl Bromide
Formula:
BrH₂CCH₂CH₃ Boiling Pt: 71 °C Melting Pt: –110 °C Density: 1.36 (20 °C) Flash Pt: 25 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000254 CAS Number: 106-94-5 EINECS: 203-445-0 UN: 2344 ADR: 3,II Merck Index: 12,08030 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...