4-Bromophenyl phenyl ether (BDE No. 3) ≥98.0%
Supplier: TCI America
Synonyms:
4-Bromodiphenyl ether, 1-Bromo-4-phenoxybenzene
B0637-500G
B0637-25G
TCB0637-500GEA
502
USD
TCB0637-500G
TCB0637-025G
4-Bromophenyl phenyl ether (BDE No. 3) ≥98.0%
4-Bromophenyl phenyl ether (BDE No. 3)
CAS Number: 101-55-3
MDL Number: MFCD00000094
Molecular Formula: C12H9BrO
Molecular Weight: 249.11
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 305
Flash Point (°C): 110
Freezing point (°C): 18
Specific Gravity (20/20): 1.43
Other Product Names / Synonyms: 4-Bromophenyl Phenyl Ether
Formula:
C₁₂H₉BrO Boiling Pt: 305 °C Density: 1.43 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00000094 CAS Number: 101-55-3 EINECS: 202-952-4 UN: 3082 ADR: 9,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...