1-Bromooctane ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1-Octyl bromide
B0626-500ML
B0626-25ML
TCB0626-500MLEA
52.88
USD
TCB0626-500ML
TCB0626-025ML
1-Bromooctane ≥98.0%
1-Bromooctane
CAS Number: 111-83-1
MDL Number: MFCD00000276
Molecular Formula: C8H17Br
Molecular Weight: 193.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 201
Flash Point (°C): 92
Specific Gravity (20/20): 1.11
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: n-Octyl Bromide
Formula:
BrH₂C(CH₂)₆CH₃ MW: 193.13 g/mol Boiling Pt: 201 °C Melting Pt: –55 °C Density: 1.11 (20 °C) Flash Pt: 92 °C Storage Temperature: Freezer |
MDL Number:
MFCD00000276 CAS Number: 111-83-1 EINECS: 203-912-9 UN: 3082 ADR: 9,III Merck Index: 12,06861 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...