1-Bromo-2-nitrobenzene ≥99.0%
Supplier: TCI America
Synonyms:
2-Bromonitrobenzene
B0622-25G
B0622-100G
TCB0622-25GEA
65
USD
TCB0622-25G
TCB0622-100G
1-Bromo-2-nitrobenzene ≥99.0%
1-Bromo-2-nitrobenzene
CAS Number: 577-19-5
MDL Number: MFCD00007045
Molecular Formula: C6H4BrNO2
Molecular Weight: 202.01
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 264
Melting point (°C): 41
Flash Point (°C): 110
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 2-Bromonitrobenzene
Formula:
C₆H₄BrNO₂ MW: 202.01 g/mol Boiling Pt: 264 °C Melting Pt: 41 °C Density: 1.62 g/cm³ (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007045 CAS Number: 577-19-5 EINECS: 209-409-0 UN: 3459 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...