1-Bromohexadecane ≥96.0%

Supplier: TCI America

B0599-25G B0599-500G B0599-100G
TCB0599-025GEA 17 USD
TCB0599-025G TCB0599-500G TCB0599-100G
1-Bromohexadecane ≥96.0%
1-Bromohexadecane

CAS Number: 112-82-3
MDL Number: MFCD00000230
Molecular Formula: C16H33Br
Molecular Weight: 305.34
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 197
Flash Point (°C): 181
Freezing point (°C): 16
Specific Gravity (20/20): 1.00


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Cetyl Bromide, Hexadecyl Bromide

Formula: BrH₂C(CH₂)₁₄CH₃
MW: 305.34 g/mol
Boiling Pt: 197 °C
Melting Pt: 14 °C
Density: 1.00 (20 °C)
Flash Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00000230
CAS Number: 112-82-3
EINECS: 204-008-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=355177612791b474a8c718986b707759bd7f6047&vid=91481c2ee13b1de9e1d7d7309a3024d4eb4a498f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR