2-Bromoethanol ≥95.0%

Supplier: TCI America
Danger

B0590-500G B0590-25G B0590-100G
TCB0590-500GEA 188 USD
TCB0590-500G TCB0590-25G TCB0590-100G
2-Bromoethanol ≥95.0%
2-Bromoethanol

CAS Number: 540-51-2
MDL Number: MFCD00002827
Molecular Formula: C2H5BrO
Molecular Weight: 124.97
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Melting point (°C): -80
Flash Point (°C): 107
Specific Gravity (20/20): 1.75
Storage Temperature: <0°C


Other Product Names / Synonyms: Ethylene Bromohydrin

Formula: BrH₂CCH₂OH
Boiling Pt: 150 °C
Melting Pt: –80 °C
Density: 1.75 (20 °C)
Flash Pt: 107 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002827
CAS Number: 540-51-2
EINECS: 208-748-1
UN: 2927
ADR: 6.1,II
Merck Index: 13,03826
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a087d25ab90921c1fe4263b06d39202518ad359&vid=2a087d25ab90921c1fe4263b06d39202518ad359&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR