1-Bromo-2-chloroethane ≥98.0%

Supplier: TCI America

Synonyms: Ethylene chlorobromide

B0572-500G B0572-25G B0572-100G
TCB0572-500GEA 287 USD
TCB0572-500G TCB0572-25G TCB0572-100G
1-Bromo-2-chloroethane ≥98.0%
1-Bromo-2-chloroethane

CAS Number: 107-04-0
MDL Number: MFCD00000962
Molecular Formula: C2H4BrCl
Molecular Weight: 143.41
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 106
Freezing point (°C): -17
Specific Gravity (20/20): 1.73


Other Product Names / Synonyms: 2-Bromoethyl Chloride, 2-Chloroethyl Bromide

Formula: BrH₂CCH₂Cl
MW: 143.41 g/mol
Boiling Pt: 106 °C
Melting Pt: –17 °C
Density: 1.73 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000962
CAS Number: 107-04-0
EINECS: 203-456-0
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a75cad09a09a3e35603312b52146b3d3e245f99&vid=b385a273fc365491a3b64ebd80a1f848544a4eef&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR