1-Bromobutane ≥98.0%
Supplier: TCI America
|
Danger
|
Synonyms:
Butyl bromide
B0560-500G
B0560-25G
TCB0560-500GEA
40.8
USD
TCB0560-500G
TCB0560-25G
1-Bromobutane ≥98.0%
1-Bromobutane
CAS Number: 109-65-9
MDL Number: MFCD00000260
Molecular Formula: C4H9Br
Molecular Weight: 137.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 101
Melting point (°C): -112
Flash Point (°C): 13
Specific Gravity (20/20): 1.28
Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Butyl Bromide
Formula:
BrH₂C(CH₂)₂CH₃ MW: 137.02 g/mol Boiling Pt: 101 °C Melting Pt: –112 °C Density: 1.28 (20 °C) Flash Pt: 13 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000260 CAS Number: 109-65-9 EINECS: 203-691-9 UN: 1126 ADR: 3 II Merck Index: 13,01552 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...