3-Bromobenzaldehyde ≥98.0%

Supplier: TCI America
Warning

B0548-500G B0548-25G B0548-100G
TCB0548-500GEA 255 USD
TCB0548-500G TCB0548-025G TCB0548-100G
3-Bromobenzaldehyde ≥98.0%
3-Bromobenzaldehyde

CAS Number: 3132-99-8
MDL Number: MFCD00003345
Molecular Formula: C7H5BrO
Molecular Weight: 185.02
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 236
Flash Point (°C): 96
Freezing point (°C): 18
Specific Gravity (20/20): 1.58

Formula: C₇H₅BrO
Boiling Pt: 236 °C
Melting Pt: 18…21 °C
Density: 1.58 (20 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00003345
CAS Number: 3132-99-8
EINECS: 221-526-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cde371501a7307ba955e90630b8f59ca7ace484&vid=184fd21bf7cee5b25ce71a197cd762bd41d3b701&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR