Methyl bromoacetate ≥98.0%

Supplier: TCI America

Synonyms: Bromoacetic methyl ester

B0533-25G B0533-100G
TCB0533-25GEA 42.27 USD
TCB0533-25G TCB0533-100G
Methyl bromoacetate ≥98.0%
Methyl bromoacetate

CAS Number: 96-32-2
MDL Number: MFCD00000189
Molecular Formula: C3H5BrO2
Molecular Weight: 152.98
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Flash Point (°C): 63
Specific Gravity (20/20): 1.66


Product Categories: Materials Chemistry, Precise Polymerization Reagents
Other Product Names / Synonyms: Bromoacetic Acid Methyl Ester

Formula: BrCH₂COOCH₃
Boiling Pt: 144 °C
Melting Pt: –50 °C
Density: 1.66 (20 °C)
Flash Pt: 63 °C
MDL Number: MFCD00000189
CAS Number: 96-32-2
EINECS: 202-499-2
UN: 2643
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdf234cb7f0f8036e5b29c7a0eb8f33a1b674b6e&vid=bdf234cb7f0f8036e5b29c7a0eb8f33a1b674b6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR