Triisopropyl borate ≥98.0%

Supplier: TCI America

Synonyms: Tri-i-propylborate

B0521-500ML B0521-25ML B0521-100ML
TCB0521-500MLEA 100 USD
TCB0521-500ML TCB0521-25ML TCB0521-100ML
Triisopropyl borate ≥98.0%
Triisopropyl borate

CAS Number: 5419-55-6
MDL Number: MFCD00008872
Molecular Formula: C9H21BO3
Molecular Weight: 188.07
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 140
Flash Point (°C): 28
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: Boric Acid Triisopropyl Ester, Isopropyl Borate

Formula: C₉H₂₁BO₃
Boiling Pt: 140 °C
Melting Pt: –59 °C
Density: 0.82 (20 °C)
Flash Pt: 28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008872
CAS Number: 5419-55-6
EINECS: 226-529-9
UN: 2616
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bfa33eb229896e917e010b7627a67e49f116510d&vid=bfa33eb229896e917e010b7627a67e49f116510d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR