N,O-Bis(trimethylsilyl)acetamide ≥80.0% (by GC)
Supplier: TCI America
|
Danger
|
Synonyms:
BSA, Silyl Derivatisation, Trimethylsilyl N-(trimethylsilyl)acetimidate, Trimethylsilyl N-trimethylsilylacetamidate
B0511-100ML
B0511-10ML
TCB0511-100MLEA
84
USD
TCB0511-100ML
TCB0511-10ML
N,O-Bis(trimethylsilyl)acetamide ≥80.0% (by GC)
N,O-Bis(trimethylsilyl)acetamide
Boiling point (°C): 73
Melting point (°C): -24
Flash Point (°C): 42
Product Categories: Synthetic Organic Chemistry, Nucleic Acid Synthesis, Silicon Compounds (for Synthesis), Analytical Science, Silylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: BSA
Formula:
C₈H₂₁NOSi₂ MW: 203.43 g/mol Boiling Pt: 73 °C Melting Pt: –24 °C Density: 0.82 g/cm³ (20 °C) Flash Pt: 42 °C |
MDL Number:
MFCD00008270 CAS Number: 10416-59-8 EINECS: 233-892-7 UN: 2920 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >80.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...