Bis(2-nitrophenyl) disulfide ≥98.0% (by GC, total nitrogen)
Supplier: TCI America
CAS Number: 1155-00-6
MDL Number: MFCD00007130
Molecular Formula: C12H8N2O4S2
Molecular Weight: 308.33
Purity/Analysis Method: >98.0% (GC,N)
Form: Crystal
Melting point (°C): 192
Other Product Names / Synonyms: 2,2'-Dinitrodiphenyl Disulfide, Di-2-nitrophenyl Disulfide
Formula:
C₁₂H₈N₂O₄S₂ Melting Pt: 192 °C Density: 1.625 g/cm³ (–123.15 °C) |
MDL Number:
MFCD00007130 CAS Number: 1155-00-6 EINECS: 214-581-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...