Bis(2-methoxyethyl)ether ≥99.0%

Supplier: TCI America
Danger

Synonyms: 2-Methoxyethyl ether, 1-Methoxy-2-(2-methoxyethoxy)ethane, Diethylene glycol dimethyl ether, Dimethyldigol, Diglyme

B0498-500ML B0498-25ML
TCB0498-500MLEA 77.34 USD
TCB0498-500ML TCB0498-25ML
Bis(2-methoxyethyl)ether ≥99.0%
Bis(2-methoxyethyl)ether

CAS Number: 111-96-6
MDL Number: MFCD00008503
Molecular Formula: C6H14O3
Molecular Weight: 134.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 162
Melting point (°C): -68
Flash Point (°C): 50
Specific Gravity (20/20): 0.95


Product Categories: Materials Chemistry, Resin Additives
Other Product Names / Synonyms: Bis(2-methoxyethyl) Ether, Diglyme, Dimethylcarbitol

Formula: C₆H₁₄O₃
Boiling Pt: 162 °C
Melting Pt: –68 °C
Density: 0.95 (20 °C)
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00008503
CAS Number: 111-96-6
EINECS: 203-924-4
UN: 3271
ADR: 3,III
Merck Index: 13,03187
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a33c2deed6b282fc4aa1aa92a9763cb2635a44e9&vid=a33c2deed6b282fc4aa1aa92a9763cb2635a44e9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR