Bis(2-chloroethyl)ether ≥99.0%

Supplier: TCI America

Synonyms: 2-Chloroethyl ether

B0472-500G B0472-25G
TCB0472-500GEA 63.8 USD
TCB0472-500G TCB0472-25G
Bis(2-chloroethyl)ether ≥99.0%
Bis(2-chloroethyl)ether

CAS Number: 111-44-4
MDL Number: MFCD00000975
Molecular Formula: C4H8Cl2O
Molecular Weight: 143.01
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 178
Melting point (°C): -47
Flash Point (°C): 55
Specific Gravity (20/20): 1.22


Other Product Names / Synonyms: 2,2'-Dichlorodiethyl Ether, 2-Chloroethyl Ether

Formula: C₄H₈Cl₂O
MW: 143.01 g/mol
Boiling Pt: 178
Melting Pt: -44
Density: 1.22 (20 °C)
Flash Pt: 55 °C
MDL Number: MFCD00000975
CAS Number: 111-44-4
EINECS: 203-870-1
UN: 1916
ADR: 6.1 II
Merck Index: 13,03091
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bce1d98675c4370c036f09940dd30ed6344ae581&vid=bce1d98675c4370c036f09940dd30ed6344ae581&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR