1,2-Bis(2-chloroethoxy)ethane ≥98.0%

Supplier: TCI America

B0471-500ML B0471-25ML
TCB0471-500MLEA 127.32 USD
TCB0471-500ML TCB0471-25ML
1,2-Bis(2-chloroethoxy)ethane ≥98.0%
1,2-Bis(2-chloroethoxy)ethane

CAS Number: 112-26-5
MDL Number: MFCD00000976
Molecular Formula: C6H12Cl2O2
Molecular Weight: 187.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 235
Flash Point (°C): 126
Specific Gravity (20/20): 1.20


Other Product Names / Synonyms: Di(2-chloroethyl) Cellosolve, Ethylene Glycol Bis(2-chloroethyl) Ether

Formula: ClH₂CCH₂OCH₂CH₂OCH₂CH₂Cl
MW: 187.07 g/mol
Boiling Pt: 235 °C
Melting Pt: –31.5 °C
Density: 1.20 (20 °C)
Flash Pt: 126 °C
Storage Temperature: Ambient
MDL Number: MFCD00000976
CAS Number: 112-26-5
EINECS: 203-952-7
UN: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96942123f64b2be7729b6fb13f00f9dd03b95796&vid=42027c47c0f4c430b84e9eeb05c0d322d1c8c7d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR