Benzyltriethylammonium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B0443-500G B0443-25G
TCB0443-500GEA 277 USD
TCB0443-500G TCB0443-025G
Benzyltriethylammonium bromide ≥98.0% (by titrimetric analysis)
Benzyltriethylammonium bromide

CAS Number: 5197-95-5
MDL Number: MFCD00011822
Molecular Formula: C13H22BrN
Molecular Weight: 272.23
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 195

Formula: C₁₃H₂₂BrN
MW: 272.23 g/mol
Melting Pt: 195 °C
Storage Temperature: Ambient
MDL Number: MFCD00011822
CAS Number: 5197-95-5
EINECS: 225-986-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27e1e91c85c7dd89d436d8f1972ccd4a12ad83fc&vid=27e1e91c85c7dd89d436d8f1972ccd4a12ad83fc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR