Dibenzyl ether ≥95.0%

Supplier: TCI America

Synonyms: Benzyl ether

B0418-500ML B0418-25ML
TCB0418-500MLEA 30.1 USD
TCB0418-500ML TCB0418-25ML
Dibenzyl ether ≥95.0%
Dibenzyl ether

CAS Number: 103-50-4
MDL Number: MFCD00004780
Molecular Formula: C14H14O
Molecular Weight: 198.27
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 298
Melting point (°C): 4
Flash Point (°C): 135
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: Benzyl Ether

Formula: C₆H₅CH₂OCH₂C₆H₅
MW: 198.26 g/mol
Boiling Pt: 298 °C
Melting Pt: 4 °C
Density: 1.04 (20 °C)
Flash Pt: 135 °C
MDL Number: MFCD00004780
CAS Number: 103-50-4
EINECS: 203-118-2
UN: 3082
ADR: 9,III
Merck Index: 13,01134
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=912303c3cfccdbc306203b65440a6702ae868c62&vid=912303c3cfccdbc306203b65440a6702ae868c62&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR