Benzyl bromide ≥98.0% stabilised

Supplier: TCI America
Warning

Synonyms: alpha-Bromotoluene

B0411-500G B0411-100G B0411-25G
TCB0411-500GEA 139.1 USD
TCB0411-500G TCB0411-100G TCB0411-25G
Benzyl bromide ≥98.0% stabilised
Benzyl bromide

(stabilized with Propylene Oxide)
CAS Number: 100-39-0
MDL Number: MFCD00000172
Molecular Formula: C7H7Br
Molecular Weight: 171.04
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 199
Flash Point (°C): 79
Specific Gravity (20/20): 1.44


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: alpha-Bromotoluene

Formula: (C₆H₅)CH₂Br
MW: 171.04 g/mol
Boiling Pt: 199
Melting Pt: -4
Density: 1.44 (20 °C)
Flash Pt: 79 °C
Storage Temperature: Ambient
MDL Number: MFCD00000172
CAS Number: 100-39-0
EINECS: 202-847-3
UN: 1737
ADR: 6.1 II
Merck Index: 13,01130
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=420f7780e91b6f65b9365d6f828e2b0c2e524c29&vid=1a0754abc42af01a2d711e2948800a8a3933e3bc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR