Benzylammonium chloride ≥98.0%
Supplier: TCI America
Synonyms:
Benzylamine hydrochloride
B0407-25G
B0407-500G
B0407-100G
TCB0407-025GEA
39.96
USD
TCB0407-025G
TCB0407-500G
TCB0407-100G
Benzylammonium chloride ≥98.0%
Benzylammonium chloride
CAS Number: 3287-99-8
MDL Number: MFCD00012852
Molecular Formula: C7H9N
Molecular Weight: 143.61
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 253
Formula:
C₇H₁₀ClN Melting Pt: 253 °C |
MDL Number:
MFCD00012852 CAS Number: 3287-99-8 EINECS: 221-943-6 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...