Benzylamine ≥99.0%

Supplier: TCI America
Danger

Synonyms: α-Aminotoluene

B0406-500ML B0406-25ML
TCB0406-500MLEA 80.67 USD
TCB0406-500ML TCB0406-25ML
Benzylamine ≥99.0%
Benzylamine

CAS Number: 100-46-9
MDL Number: MFCD00008106
Molecular Formula: C7H9N
Molecular Weight: 107.16
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 184
Flash Point (°C): 60
Specific Gravity (20/20): 0.98

Formula: (C₆H₅)CH₂NH₂
Boiling Pt: 184 °C
Melting Pt: 10 °C
Density: 0.98 (20 °C)
Flash Pt: 60 °C
Storage Temperature: Ambient
MDL Number: MFCD00008106
CAS Number: 100-46-9
EINECS: 202-854-1
UN: 2735
ADR: 8,II
Merck Index: 13,01127
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d966ec0c434d78128a66ba5bb2532b683b255d6&vid=5a2ef831079114cc46ed36ba351e99971aa362be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR