4-tert-Butylphenol ≥98.0%

Supplier: TCI America

B0230-25G B0230-500G
TCB0230-25GEA 22.11 USD
TCB0230-25G TCB0230-500G
4-tert-Butylphenol ≥98.0%
4-tert-Butylphenol

CAS Number: 98-54-4
MDL Number: MFCD00002367
Molecular Formula: C10H14O
Molecular Weight: 150.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 238
Melting point (°C): 99
Flash Point (°C): 113

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 238 °C
Melting Pt: 99 °C
Density: 1.017 g/cm³ (20 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00002367
CAS Number: 98-54-4
EINECS: 202-679-0
UN: 2430
ADR: 8,III
Merck Index: 13,01584
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=217ab529ca3be5be7096bc6eb1373e3ab14503f8&vid=007787e1fd4bbe9d5eb43d0052e33f151f4ce3c9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR