Trimethyl borate 63 - 65% in methanol
Supplier: TCI America
|
Danger
|
B0226-500ML
B0226-25ML
TCB0226-500MLEA
38
USD
TCB0226-500ML
TCB0226-25ML
Trimethyl borate 63 - 65% in methanol
Trimethyl borate
CAS Number: 121-43-7
MDL Number: MFCD00008346
Molecular Formula: C3H9BO3
Molecular Weight: 103.91
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 54
Flash Point (°C): -1
Specific Gravity (20/20): 0.88
Other Product Names / Synonyms: Boric Acid Trimethyl Ester, Methyl Borate
Formula:
C₃H₉BO₃ Boiling Pt: 54 °C Melting Pt: –5 °C Density: 0.88 (20 °C) Flash Pt: –1 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008346 CAS Number: 121-43-7 EINECS: 204-468-9 UN: 2416 ADR: 3,II Merck Index: 12,09842 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...