Benzothiazole ≥96.0%

Supplier: TCI America

Synonyms: Benzo[d]thiazole

B0092-25ML B0092-100ML
TCB0092-25MLEA 32 USD
TCB0092-25ML TCB0092-100ML
Benzothiazole ≥96.0%
Benzothiazole

CAS Number: 95-16-9
MDL Number: MFCD00005775
Molecular Formula: C7H5NS
Molecular Weight: 135.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 231
Melting point (°C): 2
Flash Point (°C): 107
Specific Gravity (20/20): 1.25

Formula: C₇H₅NS
MW: 135.19 g/mol
Boiling Pt: 231 °C
Melting Pt: 2 °C
Density: 1.25 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00005775
CAS Number: 95-16-9
EINECS: 202-396-2
UN: 2810
ADR: 6.1,III
Merck Index: 13,01108
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ac384398827cfa1e792561d2ef78d4789cda7147&vid=ac384398827cfa1e792561d2ef78d4789cda7147&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR