Benzophenone ≥99.0%

Supplier: TCI America
Warning

Synonyms: Diphenylmethanone, Diphenyl ketone

B0083-500G B0083-25G
TCB0083-500GEA 61.04 USD
TCB0083-500G TCB0083-025G
Benzophenone ≥99.0%
Benzophenone

CAS Number: 119-61-9
MDL Number: MFCD00003076
Molecular Formula: C13H10O
Molecular Weight: 182.22
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 305
Melting point (°C): 49
Flash Point (°C): 150


Product Categories: Materials Chemistry, Polymerization Initiators, Analytical Science, Estradiol, etc. (Environmental Endocrine Disruptors)
Other Product Names / Synonyms: Diphenyl Ketone

Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 305 °C
Melting Pt: 49 °C
Density: 1.085 g/cm³ (20 °C)
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00003076
CAS Number: 119-61-9
EINECS: 204-337-6
UN: 3077
ADR: 9,III
Merck Index: 13,01099
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba0e94b5eaa4fa93c78780c50b9f45339099884d&vid=ba0e94b5eaa4fa93c78780c50b9f45339099884d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR