Benzenesulphonohydrazide ≥98.0%
Supplier: TCI America
Synonyms:
Benzenesulphonic acid hydrazide, Benzenesulphonyl hydrazide
B0038-25G
B0038-100G
TCB0038-25GEA
28
USD
TCB0038-25G
TCB0038-100G
Benzenesulphonohydrazide ≥98.0%
Benzenesulphonohydrazide
CAS Number: 80-17-1
MDL Number: MFCD00007583
Molecular Formula: C6H8N2O2S
Molecular Weight: 172.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 105
Other Product Names / Synonyms: Benzenesulfonohydrazide, BSH
Formula:
C₆H₈N₂O₂S Melting Pt: 105 °C Density: 1.369 g/cm³ (20 °C) |
MDL Number:
MFCD00007583 CAS Number: 80-17-1 EINECS: 201-255-2 Merck Index: 12,07757 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...